Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A4 Recombinant Superclonal™ Antibody (16HCLC)

Catalog No. PI711108 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI711108 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI711108 Supplier Invitrogen™ Supplier No. 711108
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with Monkey, Horse and Cat Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

SERT is an integral membrane protein that transports the neurotransmitter serotonin from synaptic spaces into presynaptic neurons. The encoded protein terminates the action of serotonin and recycles it in a sodium-dependent manner. This protein is a target of psychomotor stimulants, such as amphetamines and cocaine, and is a member of the sodium:neurotransmitter symporter family. A repeat length polymorphism in the promoter of this gene has been shown to affect the rate of serotonin uptake and may play a role in sudden infant death syndrome, aggressive behavior in Alzheimer disease patients, and depression-susceptibility in people experiencing emotional trauma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A4
Applications Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Clone 16HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene SLC6A4
Gene Accession No. P31645, P31652
Gene Alias 5 OCD1; 5HT transporter; 5HTT; 5-HTT; 5-HTTLPR; 5-hydroxytryptamine (serotonin) transporter; AI323329; HGNC:11050; hSERT; HTT; Na+/Cl- dependent serotonin transporter; OCD1; OTTHUMP00000163633; serotonin transporter; serotonin transporter 1; Sert; SERT1; Slc6a4; Sodium-dependent serotonin transporter; Sodium-dependent serotonin transporter (5HT transporter) (5HTT); solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); Solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); solute carrier family 6 (neurotransmitter transporter), member 4; solute carrier family 6 (neurotransmitter transporter, serotonin), member 4; solute carrier family 6 member 4; solute carrier family 6, member 4
Gene Symbols SLC6A4
Host Species Rabbit
Immunogen Peptides corresponding to Human SLC6A4 (aa 444-462, 16-29).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25553, 6532
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.