Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC7A11 Recombinant Human Monoclonal Antibody (23H6)
GREENER_CHOICE

Catalog No. FEMA550216
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550216 Supplier Invitrogen™ Supplier No. MA550216
Only null left
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

SLC7A11 Recombinant Monoclonal Antibody for ELISA

SLC7A11 (also known as XCT) is a member of a hetromeric, sodium-independent, anionic amino acid transport system. It is highly specific for cysteine and glutamate. SLC7A11 is a mediator of Kaposi sarcoma-associated herpes virus fusion and entry into cells. Increased expression of this protein in primary gliomas is associated with neuronal cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A11
Applications ELISA, Neutralization
Classification Recombinant Monoclonal
Clone 23H6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene SLC7A11
Gene Accession No. Q9UPY5
Gene Alias 9930009M05Rik; AI451155; Amino acid transport system xc-; calcium channel blocker resistance protein CCBR1; CCBR1; cysteine/glutamate transporter; cystine/glutamate transporter; SLC7A11; sodium independent anionic amino acid transport system; solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11; solute carrier family 7 (cationic amino acid transporter, y+ system), member 11; solute carrier family 7 member 11; solute carrier family 7, (cationic amino acid transporter, y+ system) member 11; sut; xCT
Gene Symbols SLC7A11
Host Species Human
Immunogen Recombinant Human SLC7A11 protein.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23657
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.