Learn More
Invitrogen™ SLC7A4 Polyclonal Antibody

Description
Antibody detects endogenous levels of total SLC7A4.
Specifications
Specifications
| Antigen | SLC7A4 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SLC7A4 |
| Gene Accession No. | O43246 |
| Gene Alias | CAT4; CAT-4; Cationic amino acid transporter 4; HCAT3; Ig heavy chain variable region; SLC7A4; solute carrier family 7 (cationic amino acid transporter, y+ system), member 4; solute carrier family 7 (orphan transporter), member 4; solute carrier family 7 member 4; solute carrier family 7, member 4; VH; VH 3 family |
| Gene Symbols | SLC7A4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.