Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC7A5 Recombinant Rabbit Monoclonal Antibody (HL2353)
GREENER_CHOICE

Catalog No. FEMA550500 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550500 Supplier Invitrogen™ Supplier No. MA550500
  • $557.00
    $494.65 / Each of 1

    Save $62.35

    11% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

SLC7A5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Solute carrier family 7 member 5 also known as large neutral amino acids transporter small subunit 1 is a protein that in humans is encoded by the SLC7A5 gene.It is sodium-independent, high-affinity transport of large neutral amino acids such as phenylalanine, tyrosine, leucine, arginine and tryptophan, when associated with SLC3A2/4F2hc.It involved in cellular amino acid uptake and the transport of L-DOPA across the blood-brain barrier, and that of thyroid hormones triiodothyronine (T3) and thyroxine (T4) across the cell membrane in tissues such as placenta.Acts as an amino acid exchanger and plays a role in neuronal cell proliferation (neurogenesis) in brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2353
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SLC7A5
Gene Accession No. Q01650, Q63016, Q9Z127
Gene Alias 4F2; 4F2 LC; 4F2 light chain; 4F2HC; 4F2LC; 4T2HC; CD98; CD98 light chain; CD98HC; CD98LC; D0H16S474E; D16S469E; E16; hLAT1; integral membrane protein E16; large neutral amino acids transporter 1; large neutral amino acids transporter small subunit 1; LAT1; LAT-1; L-type amino acid transporter 1; MDU1; MP hLAT1; MPE16; NACAE; Protein TA1; SLC7A5; sodium-independent neutral amino acid transporter LAT1; solute carrier family 7 (amino acid transporter light chain, L system), member 5; solute carrier family 7 (cationic amino acid transporter, y+ system), member 5; solute carrier family 7 member 5; Ta1; tumor-associated protein 1; y+ system cationic amino acid transporter
Gene Symbols SLC7A5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human LAT1/SLC7A5. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20539, 50719, 8140
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.