Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104161
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104161 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104161 Supplier Invitrogen™ Supplier No. PA5104161
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC9A11.

SLC9A11 is involved in pH regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9A11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Slc9c2
Gene Accession No. Q5TAH2
Gene Alias Na(+)/H(+) exchanger 11; NHE-11; RGD1565820; SLC9A11; SLC9C2; Sodium/hydrogen exchanger 11; Solute carrier family 9 member 11; Solute carrier family 9 member C2; solute carrier family 9 member C2 (putative); solute carrier family 9, isoform 11; solute carrier family 9, member 11; solute carrier family 9, member C2 (putative)
Gene Symbols Slc9c2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC9C2(Accession Q5TAH2), corresponding to amino acid residues D479-G529.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284525, 364029
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.