Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9A8 Monoclonal Antibody (7A11)
GREENER_CHOICE

Catalog No. MA146357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA146357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA146357 Supplier Invitrogen™ Supplier No. MA146357
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Do not boil samples prior to Western Blot. Suggested positive control: antigen standard for SLC9A8 (transient overexpression lysate), HeLa whole cell lysate..

Sodium-hydrogen exchangers (NHEs), such as SLC9A8, are integral transmembrane proteins that exchange extracellular Na+ for intracellular H+. NHEs have multiple functions, including intracellular pH homeostasis, cell volume regulation, and electroneutral NaCl absorption in epithelia (Xu et al., 2008).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9A8
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7A11
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% sodium azide
Gene SLC9A8
Gene Accession No. Q4L208, Q8R4D1, Q9Y2E8
Gene Alias 1200006P13Rik; 6430709P13Rik; AI182282; DKFZp686C03237; fj61b02; FLJ42500; hypothetical protein LOC494043; KIAA0939; MGC138418; Na(+)/H(+) exchanger 8; Nhe8; NHE-8; NHE-8 antibody; SLC9A8; SLC9A8 antibody; sodium hydrogen exchanger 8; sodium/hydrogen exchanger 8; solute carrier family 9 (sodium/hydrogen exchanger), isoform 8; solute carrier family 9 (sodium/hydrogen exchanger), member 8; solute carrier family 9 member 8; solute carrier family 9 member A8; solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8; wu:fj61b02; zgc:103660
Gene Symbols SLC9A8
Host Species Mouse
Immunogen A fusion protein containing the C-terminal 89 residues of human NHE8.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23315, 311651, 77031
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.