Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542524 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542524 Supplier Invitrogen™ Supplier No. PA542524
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: INYQEQASSP CSPPARLGLD QKASPQTPGK ENIYEGDLGL GGYELKLEQT Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 93%; Rat: 100%.

This gene encodes a sodium hydrogen exchanger that is a member of the solute carrier 9 protein family. The encoded protein localizes the to the late recycling endosomes and may play an important role in maintaining cation homeostasis. Defects in this gene are associated with attention-deficit/hyperactivity disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9A9
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC9A9
Gene Accession No. Q8IVB4
Gene Alias 5730527A11Rik; 9930105B05; AI854429; AUTS16; FLJ35613; Na(+)/H(+) exchanger 9; Nbla00118; NHE9; NHE-9; putative protein product of Nbla00118; RGD1560736; Slc9a9; Sodium/hydrogen exchanger 9; sodium/proton exchanger NHE9; solute carrier family 9 (sodium/hydrogen exchanger), isoform 9; solute carrier family 9 (sodium/hydrogen exchanger), member 9; solute carrier family 9 member 9; solute carrier family 9 member A9; solute carrier family 9, subfamily A (NHE9, cation proton antiporter 9), member 9
Gene Symbols SLC9A9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SLC9A9 (aa 589-638).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285195
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.