Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLCO3A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584720
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584720 Supplier Invitrogen™ Supplier No. PA584720
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPEFLTHQYK YEAGEIRWGA EGRDVCAANG SGGDEGPDPD LICRNRTATN M Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.

SLCO3A1 mediates the Na+-independent transport of organic anions such as estrone-3-sulfate. It mediates transport of prostaglandins (PG) E1 and E2, thyroxine (T4), deltorphin II, BQ-123 and vasopressin, but not DPDPE (a derivative of enkephalin lacking an N-terminal tyrosine residue), estrone-3-sulfate, taurocholate, digoxin nor DHEAS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLCO3A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLCO3A1
Gene Accession No. Q9UIG8
Gene Alias 5830414C08Rik; androgen regulated protein 1; Anr1; FLJ40478; LOW QUALITY PROTEIN: solute carrier organic anion transporter family member 3A1; MJAM; OATP3A1; OATPD; OATP-D; OATPRP3; OATP-RP3; Organic anion transporter polypeptide-related protein 3; organic anion transporting polypeptide 3a1; organic anion-transporting polypeptide D; PGE1 transporter; Pgt2; Prostaglandin transporter subtype 2; R75096; si:ch211-196l7.1; SLC21A11; Slco3a1; Sodium-independent organic anion transporter D; solute carrier family 21 (organic anion transporter), member 11; solute carrier family 21 member 11; Solute carrier organic anion transporter family member 3A1; solute carrier organic anion transporter family, member 3A1; wu:fa93e11; zOatp3a2
Gene Symbols SLCO3A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLCO3A1. Recombinant protein control fragment (Product #RP-108514).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28232
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.