Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLCO5A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569280 Supplier Invitrogen™ Supplier No. PA569280
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

This gene encodes a 12 transmembrane domain protein that is a member of the solute carrier organic anion transporter superfamily. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLCO5A1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLCO5A1
Gene Accession No. Q9H2Y9
Gene Alias 4732454B15; A630033C23Rik; AA407151; LOW QUALITY PROTEIN: solute carrier organic anion transporter family member 5A1; OATP5A1; OATPJ; OATP-J; OATPRP4; OATP-RP4; organic anion transporter polypeptide-related protein 4; SLC21A15; SLCO5A1; solute carrier family 21 (organic anion transporter), member 15; solute carrier family 21 member 15; Solute carrier organic anion transporter family member 5A1; solute carrier organic anion transporter family, member 5A1
Gene Symbols SLCO5A1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SLCO5A1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81796
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.