Description
- Designed to easily insert into the chambers bath
- Made from 316 stainless steel with Lycra ™ threads, finished with a plastic coating
- Plastic anchors are also available for the RC-27 L family of chambers
- Slice anchors are used to secure tissue slices in a Warner recording chamber during perfusion
- While most anchors are made from stainless steel, we also have plastic anchors available for the RC-27 L chamber family. Anchors threads are made from 100 μm thick Lycra ™ at 1.0 mm, 1.5 mm, and 2.0 mm spacing.
- Kits containing two or three harps, and Nylon ™ support meshes are available for most chambers. All kits come with mesh in two sizes (112 micron and 160 micron) that may be cut and used in conjunction with the slice anchor.
Specifications
Specifications
| For Use With (Equipment) | RC-41 |
| Dimensions | 1.5 mm Thread Spacing |