Learn More
Description
- Secures tissue slices in Warner recording chambers during perfusion
- Designed for easy insertion into the chamber bath
- Made from 316 stainless steel with a plastic coating and 100 μm Lycra threads
- Thread spacing options: 1.0 mm, 1.5 mm, and 2.0 mm
- Plastic anchors available for RC-27L chamber models
- Kits available with two or three harps and Nylon support meshes
- Includes 112 μm and 160 μm mesh that can be cut for use with the anchor
Specifications
Specifications
| Type | Harp |
| For Use With (Equipment) | RC-26G Chamber |
| Dimensions | 2 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.