Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Slide-A-Lyzer™ MINI Dialysis Devices, 10K MWCO
GSA_VA

Catalog No. PI69572
Change view
Click to view available options
Product Type:
Dialysis Unit
Dialysis Unit, 15 mL Conical Tube
Dialysis Unit, 50 mL Conical Tube
Dialysis Unit, Float Buoy
Dialysis Unit, Microcentrifuge Tubes
Quantity:
1 Kit (10 Devices)
25 Devices
250 Devices
50 Devices
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
PI69572 Dialysis Unit 250 Devices
PI69570 Dialysis Unit 50 Devices
PI69574 Dialysis Unit, Microcentrifuge Tubes 1 Kit (10 Devices)
PI69576 Dialysis Unit, Float Buoy 1 Kit (10 Devices)
PI88401 Dialysis Unit, 15 mL Conical Tube 25 Devices
PI88404 Dialysis Unit, 50 mL Conical Tube 25 Devices
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. PI69572 Supplier Thermo Scientific™ Supplier No. 69572
Add to Cart
Add to Cart

Perform sample buffer exchange with these 10,000-MW cut-off (MWCO) disposable polypropylene cups with integrated, low-binding membranes.

Slide-A-Lyzer MINI Dialysis Devices, 10K MWCO, are disposable polypropylene cups with integrated, low-binding membranes for dialysis and high recovery of proteins and macromolecules greater than 10 kDa in volumes from 10 μL to 2 mL.

Slide-A-Lyzer MINI Dialysis Devices were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices. Slide-A-Lyzer MINI Dialysis Devices are manufactured in a HEPA filter/clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use.

Features of Slide-A-Lyzer MINI Dialysis Devices, 10K MWCO:
Three convenient sizes—dialysis devices for 0.1, 0.5, and 2 mL samples fit standard 1.5-mL micro-centrifuge, 15- and 50-mL conical tubes, respectively
Excellent sample recoveries—low-binding plastic and small membrane surface area minimize sample loss compared to filtration and resin systems
One-step protocol—pipette sample into the MINI device and place in tube containing the dialysis buffer; no laborious assembly, device preparation, or expensive equipment are required
100% leak-tested—innovative design does not permit 'wicking' that can occur in home-made devices
Time savings—performs desalting of nucleic acids and proteins (>95% recovery) in less than 15 minutes, which improves sample resolution, quantification, and conjugation

These 10K MWCO dialysis devices are available in three sizes (0.1, 0.5, and 2 mL capacities), and are designed to allow easy sample addition and removal using a standard laboratory pipette. The MINI devices are effective and convenient for accomplishing typical sample desalting or buffer exchange in 4–8 hours. Unlike other small sample separation devices, there is no equipment to assemble, no need for syringe adapters, and no laborious steps necessary to manipulate the small volume sample. The regenerated cellulose membrane is compatible with a number of common chemicals and buffers.

The 0.1 mL MINI dialysis devices are designed to be capped (caps included) and inserted into standard 1.5 mL micro-centrifuge tubes. Alternatively, as many as 25 MINI devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple samples inside a large beaker. The 0.5- and 2-mL MINI dialysis devices fit in and are capped by inserting them into the included 15- and 50-mL conical tubes, respectively. The tubes can be securely placed in standard laboratory racks or shakers; this feature saves space, minimizes the risk of spills or contamination, and eliminates the need for floats or large beakers of buffer. Using these MINI dialysis devices, low molecular weight contaminant removal and buffer exchange and desalting can be accomplished within 4–8 hours with efficiencies, rates, and recoveries very similar to conventional dialysis using a large dialysate volume.

Using the 0.1 mL MINI device, 100 μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes. As with all dialysis applications, the observed rate of dialysis for each sample used with these devices depends on a variety of factors including membrane to sample volume; temperature and agitation rates; the molecular weight, shape, degree of hydration, and charge of the molecules passing through the membranes; and the pH, ionic strength, and solvent characteristic of the buffers being used.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Dialysis Unit
Product Line Slide-A-Lyzer
MWCO 10 kDa
Membrane Cellulose Membrane, Dialysis
Format MINI Dialysis Device
Purification Target Buffer Exchange, Protein
Sample Volume (Metric) 0.1 mL
Kit Contents Dialysis Units
Quantity 250 Devices
Content And Storage Store at room temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.