Learn More
Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 20K MWCO
Description
Slide-A-Lyzer G2 Dialysis Cassettes (20K MWCO) provide maximum convenience, flexibility, and performance for sample dialysis.
The 20K molecular-weight cut off (MWCO) cassettes use a cellulose membrane that is rated to completely retain proteins and other macromolecules larger than 20,000 Daltons while allowing removal of buffer salts and small contaminants. The cassette membrane is composed of low-binding regenerated cellulose. Membranes and cassettes are manufactured using clean-room conditions to ensure they are contaminant-free.
Features of Slide-A-Lyzer G2 Dialysis Cassettes (20K MWCO):
• Easy-loading—pipette-accessible for easy sample loading and retrieval
• Self-floating—integrated air chambers eliminate the need for float buoys
• Sturdy construction—ensures the highest possible sample integrity and protection
• Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
• Multiple sizes—cassette capacities to optimally match 0.5 to 70 mL sample volumes
• Versatile—ideal for removing low-molecular weight contaminants, performing buffer exchange and desalting
Volume ranges:
• 0.5 mL = 0.25 to 0.75 mL
• 3 mL = 1 to 3 mL
• 15 mL = 5 to 15 mL
• 30 mL = 10 to 30 mL
• 70 mL = 25 to 70 mL
Order Info
Important Note: SKU #87736 will be discontinued on June 1, 2023, and replaced by A52977, a new generation of cassettes.Consider switching to A52977.
Specifications
Specifications
| Content And Storage | Store at room temperature |
| Purification Target | Buffer Exchange, Protein |
| Membrane | Cellulose Membrane, Dialysis |
| Format | Cassette |
| MWCO | 20 kDa |
| Product Line | Slide-A-Lyzer |
| Quantity | 8 Cassettes |
| Sample Volume (Metric) | 15 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.