Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 3.5K MWCO

Catalog No. PI87724
Change view
Click to view available options
Sample Volume (Metric):
0.5 mL
3 mL
15 mL
30 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Volume (Metric)
PI87724 15 mL
PI87722 0.5 mL
PI87723 3 mL
PI87725 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI87724 Supplier Thermo Scientific™ Supplier No. 87724

Important Note: These G2 Dialysis Cassettes will be discontinued on June 1, 2023 and replaced by G3 Dialysis Cassettes, a new generation of cassettes. Consider switching to G3 Dialysis Cassettes. Remove salts and contaminants while maximizing sample recovery with these second-generation self-floating protein cassettes.

Slide-A-Lyzer G2 Dialysis Cassettes provide maximum convenience, flexibility, and performance for sample dialysis. These 3.5K molecular weight cut off (3.5K MWCO) cassettes use a cellulose membrane to retain proteins and other macromolecules larger than 3,500 Daltons while allowing removal of salts and small contaminants. The cassette membrane is composed of low-binding regenerated cellulose. The membranes and cassettes are manufactured using clean-room conditions to ensure they are contaminant-free.

Slide-A-Lyzer G2 Dialysis Cassettes dialyze sample volumes up to 70 mL. These 3.5K MWCO cassettes allow samples to be directly injected with a syringe or pipette through the unique sample port. The cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems. The sample is easily recovered through either the pipette or syringe sample ports.

Features of Slide-A-Lyzer G2 Dialysis Cassettes (3.5K MWCO):
Easy-loading—pipette-accessible for easy sample loading and retrieval
Self-floating—integrated air chambers eliminate the need for float buoys
Sturdy construction—ensures the highest possible sample integrity and protection
Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
Versatile—available for volumes of 0.5 to 70 mL

Volume ranges:
• 0.5 mL = 0.25 to 0.75 mL
• 3 mL = 1 to 3 mL
• 15 mL = 5 to 15 mL
• 30 mL = 10 to 30 mL
• 70 mL = 25 to 70 mL

Order Info

Important Note: SKU #87724 will be discontinued on June 1, 2023 and replaced by A52967, a new generation of cassettes. Consider switching to A52967.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 3.5 kDa
Product Line Slide-A-Lyzer
Quantity 8 Cassettes
Sample Volume (Metric) 15 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.