Learn More
Epredia™ Slide Baskets and Hangers for Epredia™ ClearVue™ Coverslipper
Catalog No. A79210065 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Gemini Basket, Black Slider
Gemini Basket, White Slider
Leica Basket, Black Slider
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
A79210065 Gemini Basket, White Slider
A79210064 Gemini Basket, Black Slider
A79210066 Leica Basket, Black Slider
A79210072 Leica Basket, Black Slider
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A79210065 Supplier Epredia™ Supplier No. A79210065
Add to Cart
Add to Cart
Use these slide baskets and hangers with the Epredia™ ClearVue™ Coverslipper to make it compatible with any automated slide stainer.
  • Allow equipment compatibility with a variety of different stainers
  • Can be loaded in mix-or-match fashion
  • Sold in sets of five
Specifications
Product Type Gemini Basket, White Slider
For Use With (Equipment) Epredia ClearVue Coverslipper
Quantity 5 Pk.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov