Learn More
Scytek Laboratories SlideMaster, with Humidity Chamber, Two Slide Holders, and Waste Recepticle Components
SDP
Supplier:  Scytek Laboratories E00001
Encompass_Preferred
ScyTek Laboratories, Inc. proudly introduces SlideMaster. A humidity chamber designed by a histotechnologist that eliminates the majority of individual slide handling. SlideMaster has three different parts: A humidity chamber, two individual slide holders with white contrast bars, and a waste receptacle. It has been designed so that all three components are compact and easy to use. A maximum of 20 slides can be used per unit. This is how it works. A hydrophobic barrier (PAP Pen, Catalog #LP0001) is drawn around the tissue section and slides are put in a special slide holder that secures them into place. Drops of antibody are placed on the slides and incubated. After incubation, the individual slide holders can be tilted and secured at a 45 degree angle. All 20 slides are rinsed off in one easy motion and the buffer (PBS-Tween 20, Catalog # ABA125 or TBS-Tween 20, Catalog # TBS125) drains into the waste receptacle. Slides are gently blotted (tissue paper) at the bottom portion
Catalog No. NC0838394