Learn More
Epredia™ SlideMate™ Accessories
Catalog No. B81310019 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Crossover Ethernet Cable
Keyboard
LabWriter Software
Mini PC Premium Package
Mouse
Thermal Transfer Ribbon
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
B81310019 Mouse
B81310001 LabWriter Software
B81310005 Crossover Ethernet Cable
B81310004 Keyboard
B81320041 Thermal Transfer Ribbon
B81300031P Mini PC Premium Package
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. B81310019 Supplier Epredia™ Supplier No. B81310019
Add to Cart
Add to Cart
Enhance and optimize the Epredia™ SlideMate™ Slide Printer with Epredia™ SlideMate Slide Printer Accessories.
Accessories include:
  • Collection system Collection trays (box of 4)
  • Hopper
  • Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
Specifications
Product Type Mouse
For Use With (Equipment) SlideMate™ Slide Printer