Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ SlideMate™ Laser Microscope Slides

Catalog No. 22042959 Shop All Epredia Products
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-042-959 Blue
22-042-957 White
22-042-961 Green
22-042-953 Pink
22-042-955 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-042-959 Supplier Epredia™ Supplier No. LS5041I85241
Add to Cart
Add to Cart
This item is not returnable. View return policy

Optimized for use on the SlideMate Laser Printer, these slides provide for a clear and robust printing surface.

  • Epredia SlideMate laser slides are made from extra-white glass, manufactured to high quality standards.
  • The clarity achieved during the manufacturing process helps ensure high levels of transparency to aid in accurate diagnoses.
  • SlideMate laser slides come pre-cleaned, flat with finely ground edges.
  • Each package comes wrapped to help protect against humidity and moisture.
  • The packaging is also designed to assist with loading the slides into SlideMate laser printer hoppers to prevent contamination due to handling.
  • Manufactured in the USA.
  • SlideMate laser slides are smooth, three-quarter inch white, blue, green, pink or yellow tab slides validated to work with the SlideMate laser slide printer.
  • The tab is impervious to virtually all common laboratory chemicals and reagents.

Specifications

Length (Metric) 75 mm
Precleaned Yes
Material High Quality Glass
Frosted Not Frosted
Type Microscope Slide
Edges Finely Ground
Well Count 0
No. per Pack 144 Pk.
Width (Metric) 25 mm
No. per Case 1440/Cs.
Thickness (Metric) 1 mm
Color Blue
Corner Style 90 degrees
For Use With (Application) Epredia SlideMate Laser Printer
Length (English) 3 in.
Width (English) 1 in.
Dimensions (L x W) 75 x 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.