Learn More
Description
- Manufactured in the USA.
- SlideMate slides are smooth, three-quarter inch white and colored tab slides validated to work with the SlideMate AS slide printer.
- The tab is impervious to virtually all common laboratory chemicals and reagents.
- Available in adhesion and non-adhesion formulas.
- SlideMate Plus Adhesion Slides feature adhesion coating for routine clinical applications.
- Epredia SlideMate slides are made from extra-white glass, manufactured to high quality standards.
- The clarity achieved during the manufacturing process helps ensure high levels of transparency to aid in accurate diagnoses.
- SlideMate slides come pre-cleaned, flat with finely ground edges.
- Each package comes wrapped to help protect against humidity and moisture.
- The packaging is also designed to assist with loading the slides into SlideMate printer hoppers to prevent contamination due to handling.
Specifications
Specifications
| Length (Metric) | 75 mm |
| Material | Glass |
| No. per Case | 1440/Cs. |
| Edges | Ground |
| No. per Pack | 144 Pk. |
| Width (Metric) | 25 mm |
| Thickness (Metric) | 1 mm |
| Color | Pink |
| Coating | Non-adhesion |
| Corner Style | 90° |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.