Learn More
Typenex™ Slim 3 Blood Band

Description
- BBID – Uniquely generated 8-digit BBID helps improve safety by helping prevent blood transfusion errors. We create the BBID, so you do not have to.
- Durable – Our smudge-proof material protects the patient's information for up to 3 days during normal use.
- Reattachable – Easily reattach, relocate, and resize the Slim 3 Blood Band with the exclusive and patented R3™ Reattachment System.
Specifications
Specifications
| Closure Type | Snap |
| Closure Material | POM |
| Color | Green |
| Product Type | Blood Band |
| For Use With (Equipment) | Laboratory Information System (LIS) |
| Latex Free | Yes |
| Dimensions (L x W) | 425.5 x 11.5 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.