Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Typenex™ Slim 3 Blood Band
SDP

Catalog No. 22355683
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-355-683 Blue
22-355-685 Red
22-355-684 Orange
22-355-686 Yellow
22-355-682 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22-355-683 Supplier Typenex™ Supplier No. AB0101B
Add to Cart
Add to Cart

Typenex™ Medical developed the Slim 3 with a smaller profile for patient comfort and expanded features for patient safety. The Slim 3's less-is-more design makes it simpler for clinicians and phlebotomists to ensure the right blood gets to the right patient.

  • BBID – Uniquely generated 8-digit BBID helps improve safety by helping prevent blood transfusion errors. We create the BBID, so you do not have to.
  • Durable – Our smudge-proof material protects the patient's information for up to 3 days during normal use.
  • Reattachable – Easily reattach, relocate, and resize the Slim 3 Blood Band with the exclusive and patented R3™ Reattachment System.

Specifications

Closure Type Snap
Closure Material POM
Color Blue
Product Type Blood Band
For Use With (Equipment) Laboratory Information System (LIS)
Latex Free Yes
Dimensions (L x W) 425.5 x 11.5 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.