Learn More
Simport™ Scientific Slimsette™ Biopsy Processing/Embedding Cassettes
Catalog No. 23045382 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Gray
Green
Orange
Tan
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-382 Tan
23-045-380 Orange
23-045-381 Green
23-045-383 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-045-382 Supplier Simport™ Scientific Supplier No. M5108
Add to Cart
Add to Cart
This item is not returnable. View return policy
Slimsette™ Biopsy Processing/Embedding Cassettes incorporate a unique recessed cover, a space saving feature allowing more cassettes to be stacked in automatic labeling machines and in storage cabinets.
  • Efficient fluid exchange and drainage thanks to 392 openings each measuring 1 x 1 mm.
  • Cover does not protrude above the cassette, a great space saving feature allowing more cassettes to be stacked in automatic labeling machines and in storage cabinets.
  • Easy-to-remove pre-attached disposable cover.
Specifications
Quantity 500/Box, 3 Boxes/Cs.
Packaging Type Dispenser Boxes
Writing Face Angle 45°
Color Tan
Material Acetal
Product Type Cassette