Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Sling Psychrometers
SDPSCI_ED

Catalog No. S23982 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
15 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S23982 15 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23982 Supplier United Scientific™ Supplier No. SPCRKITPK/15
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Sling Psychrometers come as a pack of 15 kits.

  • Use this simple apparatus to measure relative humidity.
  • Includes two plastic-backed Celsius thermometers, one for dry bulb measurements and the other for wet bulb measurements.
  • The thermometers mount on a wooden handle so they can be spun to obtain temperature readings.
  • Includes an activity guide with relative humidity calculation table.
  • Requires assembly.

Specifications

Grade 5+
For Use With (Application) Relative Humidity Experiments
Includes Set of 15 Thermometers with Activity Guide
Class Size 15 Students
Product Type Hygrometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.