Learn More
MilliporeSigma™ Supelco™ Slip-On Inlet Filter
Recommended for solvent inlet applications.
Supplier:  MilliporeSigma™ Supelco™ 59277
Catalog No. 11-102-0945
Description
Description
The 10 μm filter protects the HPLC system from contamination while minimizing pump cavitation problems. Compatible with the 1.5 mm I.D. tubing used with many HPLC pumps. Also fits the 2.2 mm I.D. tubing used with Varian pumps and the 3.0 mm I.D. tubing used with Waters pumps. 316 stainless steel tip and filter element connected by a KEL-F collar.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pore Size: 10 μm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.