Learn More
Thermo Scientific™ SLIPFREE Ferrules
Catalog No. 03170505 Shop All Thermo Scientific Products
Change view
Click to view available options
Material:
Vespel
Catalog No. Material
03-170-505 Vespel
1 options
Catalog No. 03-170-505 Supplier Thermo Scientific™ Supplier No. 36024
Add to Cart
Add to Cart
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
  • Vespel ferrules replace the standard Vespel ferrules supplied with SLIPFREE column connectors
  • Kel-F and PEEK are offered for applications in which Vespel is not suitable
TRUSTED_SUSTAINABILITY
Specifications
Description Vespel sealing ferrule
Material Vespel
Type Sealing Ferrule
For Use With (Equipment) SLIPFREE Connectors for HPLC Columns