Learn More
SLK Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Guinea pig_10141
Specifications
Specifications
| Antigen | SLK |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | SLK |
| Gene Accession No. | Q9H2G2 |
| Gene Alias | LOSK, STK2, bA16H23.1, se20-9, CTCL tumor antigen se20-9, Long Ste20-like Kinase, SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrose nonfermenting like kinase, SNF1 sucrose nonfermenting like kinase, STE20-like serine/threonine-protein kinase, STE20-related kinase, STE20- |
| Gene Symbols | SLK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.