Learn More
United Scientific Supplies Slotted Weight Set of 12, Aluminum
SDPSCI_ED
Catalog No. S43721 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
Set of 12
Catalog No. Quantity
S43721 Set of 12
1 options
Catalog No. S43721 Supplier United Scientific Supplies Supplier No. WSAL12
Add to Cart
Add to Cart
Slotted weight set includes a sturdy weight hanger
  • Includes a 50 g aluminum hanger and 12 aluminum weights
  • 9 x 20 g
  • 1 x 10 g
  • 2 x 5 g
Specifications
Product Type Weight Set
Material Aluminum
Weight (Metric) 454 g
Quantity Set of 12