Learn More
United Scientific™ Slotted Weight Set of 13, with Hanger and Case
SDPSCI_ED
Catalog No. S139462 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
Set of 13
Catalog No. Quantity
S139462 Set of 13
1 options
This item is not returnable. View return policy
Catalog No. S139462 Supplier United Scientific™ Supplier No. WHST13
Add to Cart
Add to Cart
This item is not returnable. View return policy
High quality weights that last long in the classroom.
Set of 13 slotted brass weights:
  • 1 x 50 g
  • 9 x 20 g
  • 1 x 10 g
  • 2 x 5 g
  • Hanger and case also included
Specifications
Product Type Weight Set
Material Brass
Weight (Metric) 5, 10, 20, 50 g
Quantity Set of 13
Warning: Cancer and reproductive harm
Prop 65