Learn More
Grainger EAGLE Salvage Drum, HDPE
Supplier:  Grainger 4RF59
Encompass_Preferred
The EAGLE Salvage Drum HDPE is a 55 gallon open head drum made of high density polyethylene plastic. It has a rolling lug cover with a 2 inch NPS threaded bung. The drum has UN certification for hazardous materials. It measures 23.4 inches diameter by 35.4 inches high and weighs 29 pounds. The drum is translucent natural in color. It is designed for storage and transportation of both hazardous and non-hazardous liquids and can be reused. The drum has a leakproof triple rolled rim and meets DOT and OSHA regulations.
Catalog No. NC2741091