Learn More
Medchemexpress LLC HY-134541 50mg , SM-102 CAS:2089251-47-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13454150MG
Encompass_Preferred
HY-134541 50mg SM-102 CAS: 2089251-47-6 SM-102 is an ionizable amino lipid that can be used for the formation of lipid nanoparticles (LNPs). SM-102 has the potential for development of lipid nanoparticles for delivery of mRNA-based vaccines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9343
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.