Learn More
R&D Systems™ SM 102
SDP
Catalog No. 890925 Shop All Novus Products
Change view
Click to view available options
Quantity:
25 mg
Catalog No. Quantity
89-092-5 25 mg
1 options
Catalog No. 89-092-5 Supplier R&D Systems™ Supplier No. 8909/25
Add to Cart
Add to Cart
A Tocris product, part of R&D Systems. Cationic ionizable lipid
SM 102 is an ionizable cationic lipid (pKa = 6.68) used in the formulation of lipid nanoparticles (LPNs) for the delivery of mRNA in vitro and in vivo. Administration of LNPs containing SM 102 induces hepatic luciferase expression in mice. SM 102 is commonly formulated with cholesterol (Cat. No. 7945), DSPC (Cat. No. 7943), and DMG-PEG 2000 (Cat. No. 7944).
For more information on LNPs and available components see our Lipid Nanoparticles page .
Specifications
Content And Storage Store at -20°C
Product Line Tocris
Quantity 25 mg