Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SM-21 maleate
GREENER_CHOICE

Catalog No. p-4785586
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-511-0 10 mg
07-515-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 07-511-0 Supplier Tocris Bioscience™ Supplier No. 0751/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Presynaptic cholinergic modulator

Potent and selective σ2 antagonist with central effects following systemic administration. Causes increased release of acetylcholine at central muscarinic synapses. Potent analgesic (efficacy comparable to morphine) and nootropic agent.

Chemical Identifiers

CAS 155059-42-0
Molecular Formula C22H28ClNO7

Specifications

Quantity 10 mg
Formula Weight 453.92
Purity >99%
Molecular Weight (g/mol) 453.916
Chemical Name or Material SM-21 maleate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.