Learn More
ACROBiosystems Recombinant Protein;Human SMAC / Diablo Protein;E.coli;200UG;DIO,DIABLO,SMAC,DFNA64,DIABLO-S,FLJ10537,FLJ25049
Supplier:  ACROBiosystems DIOH5129200UG
Encompass_Preferred
Recombinant Protein;Human SMAC / Diablo Protein;200UG;Human SMAC, His Tag (DIO-H5129) is expressed from E.coli cells. It contains AA Ala 56 - Asp 239 (Accession # NP_063940).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.7 kDa. The protein migrates as 22 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8873
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.