Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SMAD2 (Human) ELISA Kit

Catalog No. 89333657 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-657 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-657 Supplier Abnova™ Supplier No. KA5047

Use to measure quantitatively human SMAD2. Sandwich enzyme immunoassay.

SMAD2 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human SMAD2.

Specifications

Product Type ELISA Kit
Target Species Human
For Use With (Application) Quant
Quantity 1 Kit
Storage Requirements Store at 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.