Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad2 (pS465/pS467)/Smad3 (pS423/pS425) Mouse anti-Human, Mouse, R718, Clone: O72-670, BD Phosflow™

Catalog No. BDB567080 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567080 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB567080 Supplier BD Biosciences Supplier No. 567080
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Smad2 (pS465/pS467)/Smad3 (pS423/pS425) Monoclonal antibody specifically detects Smad2 (pS465/pS467)/Smad3 (pS423/pS425) in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Smad2 (pS465/pS467)/Smad3 (pS423/pS425)
Applications Flow Cytometry, Control
Classification Monoclonal
Clone O72-670
Conjugate R718
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias SMAD2, SMAD3; MADH2, MADH3; MAD homolog 2, MAD homolog 3
Host Species Mouse
Immunogen Phosphorylated Human Smad2 Peptide
Purification Method Affinity Chromatography
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.