Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMAD6 Monoclonal Antibody (5H3)
GREENER_CHOICE

Catalog No. PIMA515687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515687 Supplier Invitrogen™ Supplier No. MA515687
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15687 targets SMAD6 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15687 immunogen is purified recombinant fragment of human SMAD6 expressed in E. Coli. . MA5-15687 detects SMAD6 which has a predicted molecular weight of approximately 53kDa.

SMAD6 (mothers against decapentaplegic homolog 6) is an antagonist of signaling by TGF-beta (transforming growth factor) type 1 receptor superfamily members; has been shown to inhibit selectively BMP (bone morphogenetic proteins) signaling by competing with the co-SMAD SMAD4 for receptor-activated SMAD1. SMAD6 is an inhibitory SMAD (I-SMAD) or antagonistic SMAD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMAD6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5H3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene SMAD6
Gene Accession No. O43541
Gene Alias AOVD2; b2b390Clo; HsT17432; MAD homolog 6; mad homolog 7; Madh6; Madh7; Mothers against decapentaplegic homolog 6; mothers against decapentaplegic, drosophila, homolog of, 6; mothers against DPP homolog 6; Msmad6; SMAD; SMAD 6; SMAD family member 6; SMAD SMAD; SMAD, mothers against DPP homolog 6; SMAD6
Gene Symbols SMAD6
Host Species Mouse
Immunogen Purified recombinant fragment of human SMAD6 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4091
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.