Learn More
Santa Cruz Biotechnology Smad7 Antibody (B-8) | 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC365846
Encompass
Anti-Smad7 Antibody (B-8) is a mouse monoclonal IgG1 κ Smad7 antibody, cited in 54 publications, provided at 200 µg/ml. Raised against amino acids 319-397 of Smad7 of human origin. Anti-Smad7 Antibody (B-8) is recommended for detection of Smad7 of mouse, rat and human origin by WB, IP, IF and ELISA.
Catalog No. NC1787723
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.