Learn More
United Scientific Supplies Small Electric Van de Graaff Generator
SDPSCI_ED
Catalog No. S27875 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Application):
Generating up to 100,000 Volts
Catalog No. For Use With (Application)
S27875 Generating up to 100,000 Volts
1 options
Catalog No. S27875 Supplier United Scientific Supplies Supplier No. VANDGF1SE
Add to Cart
Add to Cart
Motor driven electrostatic generator allows for easy viewing of the moving parts
  • Generates up to 100,000 volts
  • Aluminum collector sphere and electric motor drive supported by sturdy base
  • Clear plastic housing allows for easy viewing during experiments
  • Includes an accessory kit and activity guide
Specifications
Product Type Van De Graaff Generator
For Use With (Application) Generating up to 100,000 Volts
Includes Accessory Kit, and Activity Guide