Learn More
World Precision Instrument Small Glass Circulation Reservoir
SDP
Catalog No. 502521262
Encompass
Ussing system replacement small glass reservoir
This is hand-blown borosilicate glass, with jacketed chambers for temerature control. It is available in two sizes - 5210 holds 20mL per side, and 5362 holds 10mL per side. This is useful when expensive chemicals are involved. Reservoir condenser caps prevent air bubbles and turbulence in fluid reservoirs. These are for use with the legacy Ussing system, which has been discontinued. These replacement parts are available while supplies last.
Catalog No. 50-252-1262 Supplier World Precision Instrument Supplier No. 5362
Ussing system replacement small glass reservoir
This is hand-blown borosilicate glass, with jacketed chambers for temerature control. It is available in two sizes - 5210 holds 20mL per side, and 5362 holds 10mL per side. This is useful when expensive chemicals are involved. Reservoir condenser caps prevent air bubbles and turbulence in fluid reservoirs. These are for use with the legacy Ussing system, which has been discontinued. These replacement parts are available while supplies last.