Learn More
Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates
GREENER_CHOICE
Catalog No. 07001055 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Natural
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility
07-001-055 Natural Non-sterile
07-000-915 Clear Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-001-055 Supplier Greiner Bio-One Supplier No. 784201
Add to Cart
Edge
Add to Cart
Ensures accurate pipetting, reduces dead volume. Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates are specifically designed to accommodate heat-sealing systems.
  • Non-pyrogenic
  • Conical well design with flat bottom
  • Ensures accurate pipetting
  • Reduces dead volume of well
  • Square well geometry, compatible with CapMat plate sealers
  • Accommodates heat-sealing systems
Service Plan
No
Warranty and Services
No
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Array 16 x 24
Autoclavable Not Autoclavable
Color Natural
Material Polypropylene
Volume (Metric) Well 1 to 90 μL
No. of Wells 384
Sterility Non-sterile
Total Volume (Metric) Well 107 μL (21 μL in frustum)
Well Shape Conical Bottom (V-well)
Well Design Deep well