Learn More
Invitrogen™ Smallpox Virus A27L Polyclonal Antibody

Description
This antibody cross-reacts with Variola virus. Immunogen sequence: DNEETLKQRL TNLEK.
Specifications
Specifications
| Antigen | Smallpox Virus A27L |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene Alias | VARVgp134 |
| Host Species | Rabbit |
| Immunogen | Amino acids 42-56 of the A27L protein. |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.