Learn More
Invitrogen™ Smallpox Virus B5R Polyclonal Antibody

Description
Smallpox Virus B5R Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | Smallpox Virus B5R |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.05% sodium azide |
| Host Species | Rabbit |
| Immunogen | Amino acids 180-194 of the B5R protein. |
| Purification Method | Protein G |
| Quantity | 100 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.