Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Smallpox Virus B5R Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675330
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-330 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-330 Supplier Invitrogen™ Supplier No. PA141505
  • $587.00
    $487.70 / Each of 1

    Save $99.30

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Smallpox Virus B5R Polyclonal Antibody for Western Blot

B5R is an EEV-specific Type I membrane glycoprotein essential for EEV formation. Vaccinia virus, a member of Poxviridae family, is a dsDNA enveloped virus. It consists of 4 types of virion stages among which extracellular enveloped virus (EEV) is critical for cell-to-cell and long range viral infection. Some cellular antigens are associated with EEV and these vary with the cell type used to grow the virus. B5R consists of a large ectodomain containing regions similar to SCR domains of complement regulatory proteins followed by a transmembrane region and a cytoplasmic tail. B5R is essential for efficient wrapping of IMV, actin tail formation, normal plaque size, virus virulence, and for EEV formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Smallpox Virus B5R
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Host Species Rabbit
Immunogen Amino acids 200-214 of the B5R protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.