Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7207987 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1044 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1044 Supplier Proteintech Group Inc Supplier No. 165971AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMAP2 (Small ArfGAP2) is a Protein Coding gene. Among its related pathways are Endocytosis. Gene Ontology (GO) annotations related to this gene include GTPase activator activity. An important paralog of this gene is SMAP1.

Specifications

Antigen SMAP2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SMAP2
Gene Accession No. Q7TN29, Q8WU79
Gene Alias RP1 228H13.3, small ArfGAP2, SMAP1L, SMAP2
Gene Symbols Smap2
Host Species Rabbit
Immunogen SMAP2 Fusion Protein Ag9751
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298500, 64744, 69780
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.