Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMARCA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540911 Supplier Invitrogen™ Supplier No. PA540911
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QEEGAAAAAT EATAATEKGE KKKEKNVSSF QLKLAAKAPK SEKEMDPEYE Sequence homology: Cow: 93%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 85%; Pig: 93%; Rabbit: 93%; Rat: 86%.

Members of this G protein family are characterized by the presence of conserved SNF2 domain, a helicase-like region. They also have an ATPase activity and are thought to have a significant role regulating chromatin remodeling. Although SNF2L1 has a ubiquitous expression pattern, its active form is expressed in neurons suggesting a possible role in promoting neurite outgrowth and regulating neuronal physiology. Principal Names: SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 1 (SMARCA1); SNF2L1; SNF2LB; Possible global transcription activator SNF2L1; SWI2 Official Gene Symbol: SMARCA1 Gen Bank Accession Number: NP_620604 Gene ID: 6594 (Human) Gene Map Locus: Xq25 (Human) SNF2L1, a ~123KDa protein, is a member of SWI/SNF family of proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Smarca1
Gene Accession No. P28370
Gene Alias 5730494M04Rik; ATP-dependent helicase SMARCA1; DKFZp686D1623; DNA-dependent ATPase SNF2L; FLJ41547; global transcription activator homologous sequence; hSNF2L; ISWI; nucleosome-remodeling factor subunit SNF2L; NURF140; Probable global transcription activator SNF2L1; RGD1561046; RP3-353H6.2; SMARCA1; SNF2L; SNF2L1; SNF2LB; SNF2-like 1; SNF2L-like protein; SNF2LT; sucrose nonfermenting 2-like protein 1; SWI; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1; SWI2
Gene Symbols Smarca1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SMARCA1 (aa 35-84).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6594
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.