Learn More
Specifications
Specifications
| Gene ID (Entrez) | 56916 |
| Species | Human |
| Product Line | Novus |
| Purity | Immunogen affinity purified |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | ATP-dependent helicase 1, DKFZP762K2015, EC 3.6.1, EC 3.6.4.12, HEL1, hHEL1, KIAA1122ETL1DKFZp762K2015, subfamily a, containing DEAD/H box 1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A containing DEAD/H box 1, SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin |
| Gene Symbol | SMARCAD1 |
| Quantity | 50 μL |
| Storage Buffer | 20 mM Tris-HCl, pH 7.9, 150 mM NaCl, 15% glycerol, 2 mM MgCl2, 0.2 mM EDTA, 1 mM DTT, 10 mM beta-glycerophospate, 0.4 mM PMSF, 1 mM benzamidine and 0.4 mg/mL recombinant human insulin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
