Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ SMART 3™ Colorimeter Reagent Systems
SDPSCI_ED

Catalog No. S19545D Shop All LaMotte Company Products
Change view
Click to view available options
Analytes:
Ammonia Nitrogen
Copper
Cyanide
Dissolved Oxygen
Iron
Manganese LR
Nitrite
Phosphate HR
Phosphate LR
Silica HR
Sulfate
Zinc HR
pH
Range:
0 to 0.5 ppm
0 to 0.7 ppm
0 to 0.8 ppm
0 to 10 ppm
0 to 100 ppm
0 to 3 ppm
0 to 5 ppm
0 to 6 ppm
0 to 70 ppm
0 to 75 ppm
0.05 to 1 ppm
5 to 9.5 ppm
No. of Tests:
20
25
50
100
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Analytes Range No. of Tests
S19545D Iron 0 to 6 ppm 50
S19545G Nitrite 0 to 0.8 ppm 20
S19545A Ammonia Nitrogen 0.05 to 1 ppm 25
S19545J Sulfate 0 to 100 ppm 50
S19545H Phosphate LR 0 to 3 ppm 50
S19545C Copper 0 to 6 ppm 100
S95109 Silica HR 0 to 75 ppm 50
S95010 Iron 0 to 5 ppm 50
S94995 Cyanide 0 to 0.5 ppm 50
S94999 Dissolved Oxygen 0 to 10 ppm 100
S95104 Phosphate HR 0 to 70 ppm 50
S95115 Zinc HR 0 to 3 ppm 50
S19545P pH 5 to 9.5 ppm 100
S95012 Manganese LR 0 to 0.7 ppm 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. S19545D Supplier LaMotte™ Supplier No. 3648SC
Add to Cart
Add to Cart

SMART 3™ Colorimeter Reagent Systems for use in water quality testing and environmental studies.

Choose from the following test factors:

  • Zinc Laboratory Reagent (LR)
  • Manganese
  • Aluminum
  • Ozone
  • Cyanide
  • Phosphate
  • Dissolved Oxygen
  • Chromium Hexavalent
  • Ammonia Nitrogen LR

Specifications

Analytes Iron
For Use With (Application) Water testing and environmental studies
For Use With (Equipment) SMART colorimeter
No. of Tests 50
Range 0 to 6 ppm
Test Method Bipyridyl
Type Iron Reagent System
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.