Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SMART Digest™ Pepsin Kits

Catalog No. 03254062
Change view
Click to view available options
Description:
Bulk Resin (No Collection Plate)
with Collection Plate
with Filter and Collection Plate
with Magnetic Bead, Bulk Option (No Collection Plate)
with SOLAμ Collection Plate
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
03-254-062 Bulk Resin (No Collection Plate)
03-254-059 with Collection Plate
03-254-060 with Filter and Collection Plate
03-254-061 with SOLAμ Collection Plate
03-254-063 with Magnetic Bead, Bulk Option (No Collection Plate)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-254-062 Supplier Thermo Scientific™ Supplier No. 60109110B

Use these pepsin protein digestion kits in your peptide mapping and peptide quantitation workflows to obtain high quality analytical results.

Use Thermo Scientific™ SMART Digest™ Pepsin Kits in your peptide mapping and peptide quantitation workflows to obtain high quality analytical results. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Pepsin Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.

How SMART Digest Pepsin Kits work
Pepsin cleaves proteins preferentially at carboxylic groups of aromatic amino acids such as phenylalanine, tryptophan and tyrosine. It will not cleave at bonds containing valine, alanine or glycine. Pepsin is often used to generate antibody fragments. Pepsin is used to generate F(ab')2 fragments by removing only that portion of the Fc domain beyond the hinge region. The resulting F(ab')2 fragments are composed of two antibody-binding Fab' fragments connected by disulfides in the hinge region.

TRUSTED_SUSTAINABILITY

Specifications

Description Bulk Resin (No Collection Plate)
Capture None
Cleavage Hydrophobic residues in P1 and P1' positions at the carboxyl side of phenylalanine and leucine and at the carboxyl side of glutamic acid
Enzyme Pepsin
Format Bulk Non-magnetic
For Use With (Application) Peptide Quantitation, Peptide Mapping
Product Line SMART Digest
Type Digest Pepsin Kit
Includes Bulk Non-magnetic SMART Digest Resin, SMART Digest Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.