Learn More
Broadley James Corporation SMART POLARIZER
Supplier:  Broadley James Corporation AM9660VP
Encompass
Full zero and air test to verify the dissolved oxygen sensor is fit for purpose.Prevents a faulty sensor being fitted to a vessel.Simplifies testing procedures.Reduces operational costs.
Catalog No. NC1822382
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.