Learn More
JDM SCIENTIFIC LLC Smart Rack starter set Chemistry
Supplier:  JDM SCIENTIFIC LLC 1508
Encompass_Preferred
The SmartRack Starter Set Chemistry has been specially designed for work in the field of chemistry and includes the following components (item number of the individual components in brackets for better understanding):8 components:- 1 x SmartRack (Item no. 01704)- 2 x Holder for glassware with ST14, plus closure (Item no. 01112)- 2 x Holder for glassware with ST29, plus closure (Item no. 01115)- 2 x Holder for chromatography column, Ø 25 - 35 mm (Item no. 01107)- 1 x Storage box (Item no. 01142)
Catalog No. 50-255-8513
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.